Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Proteasome Inhibitor I

Catalog No. p-7060142 Shop All MilliporeSigma Products
Click to view available options
Quantity:
1 mg
5 mg

The Proteasome Inhibitor I controls the biological activity of Proteasome. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

  • Peptide Sequence: Z-Ile-Glu(OtBu)-Ala-Leu-CHO
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color White
Target Proteins Chymotrypsin-like activity of the multicatalytic proteinase complex (MCP; 20S proteasome) in HT4 cells
Molecular Formula C32H50N4O8
Formula Weight 618.8g/mol
Solubility DMSO, EtOH, or MeOH
Form Solid
Quantity 1 mg
Purity 90% by HPLC
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.