Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ AMPK Inhibitor, Compound C
Shop All MilliporeSigma Products
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Description
- Packaged Under Inert Gas
- Cell Permeable
Specifications
Specifications
| Color | Light Yellow |
| Target Proteins | AMPK |
| CAS | 866405-64-3 |
| Molecular Formula | C24H25N5O |
| Formula Weight | 399.5g/mol |
| Solubility | DMSO, 100 mM HCl, or MeOH |
| Content And Storage | Protect from light |
| Form | Solid |
| Quantity | 1 mg |
| Purity | 95% by HPLC |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction