Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ 2-Aminopurine
Click to view available options
Quantity:
250 mg
Description
A cell permeable adenine analog that is widely used as a versatile fluorescent probe to investigate RNA and DNA secondary and tertiary structures and their conformation dynamics in response to local environment when interacting with other biomolecules with great sensitivity. Forms stable base pairs with neucleobase uracil found in RNA and thymine in DNA, and moderately stable base pairs with cytosine. Exhibits intensive fluorescence emission (lAbsmax = 303 nM, lflmax = 370 nM, and fluorescence quantum yield in solution QFI = 0.68) and longer excitation wavelength compared with other neucleobases enabling its selective excitation with low background signals.
Please note that the molecular weight for this compound is batch-specific due to variable water content.
Please note that the molecular weight for this compound is batch-specific due to variable water content.

Specifications
Specifications
Color | Yellow |
Description | 97% by HPLC |
Target Proteins | DNA and RNA |
CAS | 452-06-2 |
Molecular Formula | C5H5N5 |
Solubility | DMSO |
Content And Storage | Protect from light |
Form | Powder |
Quantity | 250 mg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction