Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ InSolution™ Q-VD-OPh, Non-O-methylated
Click to view available options
Quantity:
1 mg
5 mg
Description
- Packaged Under Inert Gas
- Peptide Sequence: Q-Val-Asp-CH2-OPh
- Cell Permeable

Specifications
Specifications
Formulation | Supplied as a 10 mM (1mg/195μl) solution in DMSO. |
Target Proteins | caspase-1 |
Molecular Formula | C26H25F2N3O6 |
Formula Weight | 513.8g/mol |
Content And Storage | Hygroscopic, Protect from light |
Form | Liquid |
Quantity | 5 mg |
Purity | 90% by HPLC |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction