Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ Calphostin C, Cladosporium cladosporioides
Shop All MilliporeSigma Products
Click to view available options
Quantity:
50 μg
Description
A cell permeable, highly specific inhibitor of protein kinase C (IC50 = 50nM) that interacts with the protein’s regulatory domain by competing at the binding site of diacylglycerol and phorbol esters. Does not compete with Ca2+ or phospholipids. At higher concentrations inhibits myosin light chain kinase (IC50 > 5μM), protein kinase A (IC50 > 50μM), protein kinase G (IC50 > 25μM), and p60v- src (IC50 > 50μM). Induces apoptotic DNA fragmentation and cell death in HL-60 human promyelocytic leukemia cells. Requires brief exposure to light for activation.

Specifications
Specifications
Color | Light Brown |
Target Proteins | PKC |
CAS | 121263-19-2 |
Molecular Formula | C44H38O14 |
Formula Weight | 790.8g/mol |
Solubility | DMSO or EtOH |
Content And Storage | Protect from light |
Form | Lyophilized |
Quantity | 50 μg |
Purity | 95% by HPLC |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction