Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Diethyl Pyrocarbonate

Catalog No. p-7060069
Click to view available options
Quantity:
25 g
100 g

Inhibits ryanodine binding to ryanodine/Ca2+ receptor channel in skeletal muscle in a dose- and time-dependent manner and increases the Ca2+ permeability of SR vesicles.

Inhibits ryanodine binding to ryanodine/Ca2+ receptor channel in skeletal muscle in a dose- and time-dependent manner and increases the Ca2+ permeability of SR vesicles. Useful for specific inactivation of nucleases during isolation of undegraded polynucleotides. Also inhibits platelet-activating factor acetyl hydrolase. Has been used as a probe for the topography of 5.8S rRNA in yeast ribosomes.
TRUSTED_SUSTAINABILITY

Specifications

Color Clear Colorless
Target Proteins Inhibits ryanodine binding to ryanodine/Ca2+ receptor channel in skeletal muscle
CAS 1609-47-8
Molecular Formula C6H10O5
Formula Weight 162.1g/mol
Solubility CHCl3 or EtOH
Content And Storage Protect from light
Form Liquid
Quantity 100 g
Purity 97% by titration
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.