Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ Lactacystin, Synthetic
Click to view available options
Quantity:
1 mg
200 μg
400 μg
Description
An irreversible proteasome inhibitor. A Streptomyces metabolite that acts as a highly specific inhibitor of the 20S proteasome (MCP; multicatalytic proteinase complex) (IC50 = 500nM for 20S proteasome). Blocks proteasome activity by targeting the catalytic β-subunit. Acts as a covalent inhibitor of the chymotrypsin- and trypsin-like activities of the proteasome. Induces neurite outgrowth in Neuro 2A mouse neuroblastoma cells and inhibits progression of synchronized Neuro 2A cells and MG-63 human osteosarcoma cells beyond the G1 phase of the cell cycle. Reported to induce apoptosis in human monoblast U937 cells. Also inhibits NF-κB activation (IC50 = 10μM).

Specifications
Specifications
Color | White |
Target Proteins | 20S proteasome (MCP; multicatalytic proteinase complex) |
CAS | 1258004-00-0 |
Molecular Formula | C15H24N2O7S |
Formula Weight | 376.4g/mol |
Solubility | DMSO or H2O |
Form | Solid |
Quantity | 200 μg |
Purity | 99% by HPLC |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction