Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ Licochalcone-A, Synthetic
Shop All MilliporeSigma Products
Click to view available options
Quantity:
10 mg
50 mg
Description
Packaged Under Inert Gas

Specifications
Specifications
Color | Yellow |
Target Proteins | Anti-tumor and antiparasitic properties |
CAS | 58749-22-7 |
Molecular Formula | C21H22O4 |
Formula Weight | 338.4g/mol |
Solubility | DMSO or EtOH |
Content And Storage | Protect from light |
Form | Solid |
Quantity | 10 mg |
Purity | 96% by HPLC |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction