Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ MEK Inhibitor I
Shop All MilliporeSigma Products
Click to view available options
Quantity:
1 mg
Description
- Packaged Under Inert Gas
- Cell Permeable

Specifications
Specifications
Color | White |
Target Proteins | MEK |
CAS | 297744-42-4 |
Molecular Formula | C21H18N4OS |
Formula Weight | 374.5g/mol |
Solubility | DMSO or MeOH |
Content And Storage | Protect from light |
Form | Solid |
Quantity | 1 mg |
Purity | 95% by HPLC (sum of two isomers) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction