Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ 2-Methoxyestradiol

Catalog No. 45418010MG Shop All MilliporeSigma Products
Click to view available options
Quantity:
10 mg
50 mg

A natural metabolite of 17β-estradiol that is devoid of estrogenic activity.

A natural metabolite of 17β-estradiol that is devoid of estrogenic activity. Inhibits cell proliferation and angiogenesis. Markedly inhibits the neovascularization of solid tumors and suppresses tumor growth in mice. Binds to the colchicine binding site of tubulin, and has been suggested to function as a natural regulator of microtubule assembly and function.
TRUSTED_SUSTAINABILITY

Specifications

Color White
Target Proteins Inhibits cell proliferation and angiogenesis
CAS 362-07-2
Molecular Formula C19H26O3
Formula Weight 302.4g/mol
Solubility DMSO or EtOH
Form Crystalline Solid
Quantity 10 mg
Purity 90% by HPLC
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.