Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ MG-132
Shop All MilliporeSigma Products
Click to view available options
Quantity:
1 mg
5 mg
10 mg
20 mg
Description
- Peptide Sequence: Z-Leu-Leu-Leu-CHO
- Cell Permeable
Specifications
Specifications
| Color | White |
| Target Proteins | proteasome |
| CAS | 133407-82-6 |
| Molecular Formula | C26H41N3O5 |
| Formula Weight | 475.6g/mol |
| Solubility | DMSO or EtOH |
| Form | Solid |
| Quantity | 1 mg |
| Purity | 98% by HPLC |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction