Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Akt Inhibitor IV

Catalog No. 1240115MG Shop All MilliporeSigma Products
Click to view available options
Quantity:
1 mg
5 mg

The Akt Inhibitor IV, also referenced under CAS 681281-88-9, controls the biological activity of Akt.

  • Packaged Under Inert Gas
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color Yellow
Target Proteins Akt
CAS 681281-88-9
Molecular Formula C31H27IN4S
Formula Weight 614.6g/mol
Solubility DMSO
Content And Storage Hygroscopic, Protect from light
Form Solid
Quantity 5 mg
Purity 98% by HPLC
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.