Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ 4α-Phorbol-12,13-didecanoate
Click to view available options
Quantity:
1 mg
5 mg
Description
Packaged Under Inert Gas

Specifications
Specifications
Target Proteins | A negative control for Phorbol-12,13-didecanoate (PDD) and Phorbol-12-myristate-13-acetate (PMA) |
CAS | 27536-56-7 |
Molecular Formula | C40H64O8 |
Formula Weight | 672.9g/mol |
Solubility | DMSO or EtOH |
Form | Solid |
Quantity | 1 mg |
Purity | >99% by HPLC |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction