Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Albumin, Human Serum, Fraction V, Low Heavy Metals

Catalog No. 1266610GM Shop All MilliporeSigma Products
Click to view available options
Quantity:
1 g
10 g
100 g

Catalog No. 12-666-10GM

Supplier: MilliporeSigma™ 1266610GM

Only null left
Add to Cart
Add to Cart

Prepared under non-denaturing conditions by modification of Cohn procedure

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Protein Standard, Protein Base or Filler, Stabilizer and Diluent in Serology and Immunohematology
Quantity 10 g
Source Prepared from serum that has been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV.
Storage Requirements 2° to 8°C
Form Solid
Purity or Quality Grade ≥95% by SDS PAGE

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.