Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Protease Inhibitor Cocktail Set I

Catalog No. p-7060135
Click to view available options
Quantity:
1 Vial
10 Vials

A cocktail of five protease inhibitors that will inhibit a broad range of proteases and esterases. Supplied with a data sheet.

A cocktail of five protease inhibitors that will inhibit a broad range of proteases and esterases. Supplied with a data sheet.
TRUSTED_SUSTAINABILITY

Specifications

Solubility H2O
Content And Storage Hygroscopic
Form Lyophilized
Quantity 1 Vial
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.