Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Protease Inhibitor Cocktail Set VI

Catalog No. p-7060137 Shop All MilliporeSigma Products
Click to view available options
Quantity:
1 Set
1 mL

The Protease Inhibitor Cocktail Set VI controls the activity of Protease. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

A cocktail of six protease inhibitors with broad specificity for the inhibition of aspartic, cysteine, serine, and metalloproteases as well as aminopeptidases. This cocktail is recommended for use with plant cell extracts. Each vial contains 200mm AEBSF, HCl (Cat. No. 101500), 10mm Bestatin (Cat. No. 200484), 3mm E-64 (Cat. No. 324890), 2mm Leupeptin Hemisulfate (Cat. No. 108975), 2mm Pepstatin A (Cat. No. 516481), and 500mm 1,10-Phenanthroline (Cat. No. 516705). OnemL is recommended for 30 g of various plant tissues. Supplied with a data sheet. Note: 1 set = 5 x 1 ml.
TRUSTED_SUSTAINABILITY

Specifications

Formulation In 1mL DMSO.
Target Proteins Aspartic, cysteine, serine, and metalloproteases as well as aminopeptidases.
Form Liquid
Quantity 1 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.