Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ Rapamycin
Click to view available options
Quantity:
1 mg
10 mg
100 μg
Description
Anti-fungal and immunosuppressant. Selectively inhibits the mammalian target of rapamycin (mTOR) and blocks the subsequent activation of p70 S6 kinase (IC50 = 50 pM). Specifically inhibits mTORC1, but mTORC2 that phosphorylates Akt at Ser473 appears to be insensitive to rapamycin. Prevents the translational activation of IGF-II. Also prevents resting T-cells from entering the cell cycle, but does not directly arrest cell cycle progression. Shown to inhibit later signaling events, such as p110Rb phosphorylation, p34cdc2 kinase activation, and cyclin A synthesis. Exhibits strong binding to FK-506 binding proteins. Also reported to induce apoptosis in a murine B-cell line, to inhibit lymphokine-induced cell proliferation at the G1 phase, and to irreversibly arrest Saccharomyces cerevisiae cells in the G1 phase. A 5mm (500μg/109μl) solution of Rapamycin (Cat. No. 553211) in DMSO and a 10mm (1mg/109μl) solution of Rapamycin (Cat. No. 553212) in EtOH is also available.

Specifications
Specifications
Color | Clear |
Target Proteins | Mammalian target of rapamycin (mTOR) |
CAS | 53123-88-9 |
Molecular Formula | C51H79NO13 |
Formula Weight | 914.2g/mol |
Solubility | DMSO or MeOH |
Form | Film |
Quantity | 100 μg |
Purity | 99% by HPLC |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction