Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Rapamycin

Catalog No. p-7060146
Click to view available options
Quantity:
1 mg

Rapamycin, CAS 53123-88-9, is an immunosuppressant that selectively inhibits mTOR and blocks the subsequent activation of p70 S6 kinase (IC50 = 50 pM).

Anti-fungal and immunosuppressant. Selectively inhibits the mammalian target of rapamycin (mTOR) and blocks the subsequent activation of p70 S6 kinase (IC50 = 50 pM). Specifically inhibits mTORC1, but mTORC2 that phosphorylates Akt at Ser473 appears to be insensitive to rapamycin. Prevents the translational activation of IGF-II. Also prevents resting T-cells from entering the cell cycle, but does not directly arrest cell cycle progression. Shown to inhibit later signaling events, such as p110Rb phosphorylation, p34cdc2 kinase activation, and cyclin A synthesis. Exhibits strong binding to FK-506 binding proteins. Also reported to induce apoptosis in a murine B-cell line, to inhibit lymphokine-induced cell proliferation at the G1 phase, and to irreversibly arrest Saccharomyces cerevisiae cells in the G1 phase. A 5mm (500μg/109μl) solution of Rapamycin (Cat. No. 553211) in DMSO and a 10mm (1mg/109μl) solution of Rapamycin (Cat. No. 553212) in EtOH is also available.
TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Target Proteins Mammalian target of rapamycin (mTOR)
CAS 53123-88-9
Molecular Formula C51H79NO13
Formula Weight 914.2g/mol
Solubility DMSO or MeOH
Form Film
Quantity 1 mg
Purity 99% by HPLC
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.