Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Rho Kinase Inhibitor

Catalog No. p-7060149 Shop All MilliporeSigma Products
Click to view available options
Quantity:
1 mg
5 mg

The Rho kinase inhibitor, CAS 872543-07-6, is a cell-permeable, highly specific, reversible, potent, and ATP-competitive inhibitor of Rho-associated kinase (ROCK; Ki = 1.6 nM).

  • Packaged Under Inert Gas
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color White
Target Proteins ROCK
Molecular Formula C16H21N3O2S·2HCl
Formula Weight 392.3g/mol
Solubility H2O or MeOH
Content And Storage Hygroscopic, Protect from light
Form Solid
Quantity 1 mg
Purity 95% by HPLC
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.