Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ AMN082 Dihydrochloride
Click to view available options
:
10mg
Description
A potent, selective and systemically active positive allosteric modulator for mGlu7 receptor (EC50 = 64-290nM). One of the most frequently used agent for assessing the functional roles of mGlu7 receptors in a variety of brain functions (i.e. sleep pattern, learning and memory, and locomotion) and disorders (i.e. depression, anxiety, and Parkinsons disease). Induces internalization of mGlu7 receptors. Also acts on NMDA, AMPA, 5-HT receptors and norepinephrine transporter.

Specifications
Specifications
Color | White |
Description | 98% by HPLC |
Target Proteins | mGlu7 receptors |
Molecular Formula | C28H28N2·2HCl |
Solubility | DMSO |
Content And Storage | Protect from light |
Form | Solid |
Quantity | 10 mg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction