Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Purmorphamine

Catalog No. p-7060144 Shop All MilliporeSigma Products
Click to view available options
Quantity:
5 mg
20 mg

A cell-permeable activator of Hedgehog signaling that induces osteoblast differentiation of multipotent mesenchymal progenitor cells C3H10T1/2 (EC50 = 1 μM).

  • Packaged Under Inert Gas
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Target Proteins Activation of the Hedgehog signaling pathway
CAS 483367-10-8
Molecular Formula C31H32N6O2
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 5 mg
Purity 98% by HPLC
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.