Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Smoothened Agonist, SAG

Catalog No. p-7060163
Click to view available options
Quantity:
1 mg
5 mg

A cell-permeable Smoothened Agonist, SAG, CAS 364590-63-6, modulates the coupling of Smo with its downstream effector by interacting with the Smo heptahelical domain (KD = 59 nM).

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color Light Yellow
CAS 364590-63-6
Molecular Formula C28H28ClN3OS·2HCl·2H2O
Formula Weight 599g/mol
Solubility DMSO or H2O
Content And Storage Hygroscopic, Protect from light
Form Solid
Quantity 1 mg
Purity 98% by HPLC
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.