Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ γ-Secretase Inhibitor IX
Shop All MilliporeSigma Products
Click to view available options
Quantity:
5 mg
10 mg
Description
- Packaged Under Inert Gas
- Cell Permeable

Specifications
Specifications
Target Proteins | Aβtotal |
CAS | 208255-80-5 |
Molecular Formula | C23H26F2N2O4 |
Formula Weight | 432.5g/mol |
Solubility | DMSO |
Content And Storage | Protect from light |
Form | Lyophilized |
Quantity | 5 mg |
Purity | 98% by HPLC |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction