Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ AMPK Inhibitor, Compound C

Catalog No. 17126010MG Shop All MilliporeSigma Products
Click to view available options
Quantity:
1 mg
5 mg
10 mg

Catalog No. 17-126-010MG

Supplier: MilliporeSigma™ 17126010MG

Only null left
Add to Cart
Add to Cart

AMPK Inhibitor, Compound C, CAS 866405-64-3, is a cell-permeable compound that inhibits KDR/VEGFR2, ALK2/BMPR-I, and AMPK kinase activities (IC50 = 25.1, 148, and 234.6 nM, respectively).

  • Packaged Under Inert Gas
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color Light Yellow
Target Proteins AMPK
CAS 866405-64-3
Molecular Formula C24H25N5O
Formula Weight 399.5g/mol
Solubility DMSO, 100 mM HCl, or MeOH
Content And Storage Protect from light
Form Solid
Quantity 10 mg
Purity 95% by HPLC
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.