Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ SU5402

Catalog No. p-7060167 Shop All MilliporeSigma Products
Click to view available options
Quantity:
1 mg
2 mg
500 μg

SU5402, CAS 215543-92-3, is a cell-permeable, reversible, and ATP-competitive inhibitor of the tyrosine kinase activity of FGFR1 (IC50 = 10 to 20 μM in the presence of 1 mM ATP).

Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Target Proteins Fibroblast growth factor receptor 1 (FGFR1)
Molecular Formula C17H16N2O3
Formula Weight 296.3g/mol
Solubility DMSO
Content And Storage Protect from light
Form Lyophilized
Quantity 500 μg
Purity 95% by HPLC
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.