Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Trypsin Inhibitor, Soybean, High Activity

Catalog No. p-7060179 Shop All MilliporeSigma Products
Click to view available options
Quantity:
100 mg
250 mg

Catalog No. 65-035-7250MG

Supplier: MilliporeSigma™ 650357250MG

Only null left
Add to Cart
Add to Cart

Trypsin Inhibitor, Soybean, High Activity, CAS 9035-81-8 is of the several protease inhibitors in soybeans. Inhibits trypsin in 1:1 stoichiometry.

Component VI of Rackis. Does not inhibit chymotrypsin activity.
TRUSTED_SUSTAINABILITY

Specifications

Color White
Target Proteins Trypsin
CAS 9035-81-8
Formula Weight 20000g/mol
Solubility 67 mM potassium phosphate buffer, pH 7.6
Content And Storage Protect from moisture
Form Lyophilized
Quantity 250 mg
Purity 90% by SDS-PAGE
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.