Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Triptolide, Tripterygium wilfordii

Catalog No. p-7060177 Shop All MilliporeSigma Products
Click to view available options
Quantity:
1 mg
5 mg

A novel diterpene triepoxide isolated from the Chinese herb Tripterygium wilfordii that acts as a potent immunosuppressant and anti-inflammatory agent.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Target Proteins Activate DEVD-cleaving caspases
CAS 38748-32-2
Molecular Formula C20H24O6
Formula Weight 360.4g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 1 mg
Purity 95% by HPLC
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.