Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ SB 203580, Hydrochloride
Shop All MilliporeSigma Products
Click to view available options
Quantity:
1 mg
10 mg
Description
Cell Permeable
Specifications
Specifications
| Color | Bright Yellow |
| Target Proteins | p38 MAP kinase |
| Molecular Formula | C21H16FN3OS·HCl |
| Formula Weight | 413.9g/mol |
| Solubility | DMSO or H2O |
| Content And Storage | Hygroscopic, Protect from light |
| Form | Solid |
| Quantity | 1 mg |
| Purity | 95% by HPLC |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction