Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ Zebularine
Shop All MilliporeSigma Products
Click to view available options
Quantity:
10 mg
25 mg
Description
- Packaged Under Inert Gas
- Cell Permeable

Specifications
Specifications
Color | Off-white |
Target Proteins | Cytidine deaminase |
CAS | 3690-10-6 |
Molecular Formula | C9H12N2O5 |
Formula Weight | 228.2g/mol |
Solubility | DMSO or MeOH |
Form | Solid |
Quantity | 25 mg |
Purity | 95% by HPLC |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction