Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Aprotinin, Bovine, Recombinant, Nicotiana sp., Animal-Free

Catalog No. 6163715MG Shop All MilliporeSigma Products
Click to view available options
Quantity:
1 mg
5 mg
25 mg

Aprotinin, Bovine, Recombinant, Nicotiana sp, Animal-Free - CAS 9087-70-1, is a heat and acid-stable competitive, reversible inhibitor of serine proteases.

Recombinant, bovine aprotinin supplied without animal-derived components. Aprotinin is a competitive, reversible inhibitor of proteolytic and esterolytic activity. A relatively heat- and acid-stable serine protease inhibitor. Forms a tight complex, blocking the active site of the enzyme. Effective at concentrations equimolar with protease. Inhibits several proteases, including coagulation factors in the prephase of blood clotting, tissue and leukocytic proteinases, chymotrypsin, trypsin (Kd = 5 x 10-14 M), plasmin (Kd = 2.3 x 10-10 M), and kallikrein (Kd = 1 x 10-7 M). Proteases not inhibited by aprotinin include Factor Xa, thrombin, pepsin, papain, and carboxypeptidases A and B. Useful for protein purification and for extending the life of cells in culture by preventing proteolytic damage.
TRUSTED_SUSTAINABILITY

Specifications

Color Clear Colorless to Amber
Formulation In 0.9% NaCl. Contains no animal-derived components.
Target Proteins chymotrypsin
CAS 9087-70-1
Molecular Formula C284H432N84O79S7
Formula Weight 6512g/mol
Form Liquid
Quantity 5 mg
Purity 95% by SDS-PAGE
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.