Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ E-64 Protease Inhibitor

Catalog No. p-7060071 Shop All MilliporeSigma Products
Click to view available options
Quantity:
1 mg
5 mg
25 mg

The E-64 Protease Inhibitor, also referenced under CAS 66701-25-5, controls the biological activity of E-64 Protease. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

Irreversible inhibitor of cysteine proteases. Interacts with the Sn subsites of proteases. Has no action on cysteine residues in other proteins. Inhibits activation-induced programmed cell death and restores defective immune responses in HIV+ donors. Specific active site titrant.
TRUSTED_SUSTAINABILITY

Specifications

Color White
Target Proteins cysteine proteases
CAS 66701-25-5
Molecular Formula C15H27N5O5
Formula Weight 357.4g/mol
Solubility DMSO
Form Solid
Quantity 1 mg
Purity 98% by HPLC
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.