Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ β-Glycerophosphate, Disodium Salt, Pentahydrate

Catalog No. p-7060076 Shop All MilliporeSigma Products
Click to view available options
Quantity:
50 g
100 g
250 g

A phosphate group donor in matrix mineralization studies that acts as a protein phosphatase inhibitor.

Contaminants: L-α-glycerophosphate: ≤5%
TRUSTED_SUSTAINABILITY

Specifications

Color White
Target Proteins protein phosphatase
CAS 13408-09-8
Molecular Formula C3H7O6PNa2·5H2O
Formula Weight 306.1g/mol
Solubility H2O
Content And Storage Protect from moisture
Form Solid
Quantity 100 g
Purity 99% by 31P NMR
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.