Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ BHQ
Click to view available options
Quantity:
100 mg
Description
Mobilizes Ca2+ specifically from Ins(1,4,5)P3-sensitive Ca2+ stores by inhibiting microsomal and sarcoplasmic reticulum Ca2+-ATPase activity. Does not affect mitochondrial Ca2+ fluxes or plasma membrane Ca2+/Mg2+ ATPase activity. Inhibits prostaglandin E2 and prostacyclin formation in osteoblast-like cell lines.

Specifications
Specifications
Color | White |
Description | 99% by HPLC |
Target Proteins | micosomal and sarcoplasmic reticulation Ca2+ ATPase activity |
CAS | 88-58-4 |
Molecular Formula | C14H22O2 |
Formula Weight | 222.3g/mol |
Solubility | DMSO, EtOH or MeOH |
Form | Solid |
Quantity | 100 mg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction