Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ Bovine Plasma Fibronectin
Shop All MilliporeSigma Products
Click to view available options
Quantity:
1 mg
5 mg
Description
EMD Millipore Calbiochem™ Bovine Plasma Fibronectin is an effective agent for promoting attachment of cells to commonly-used culture substrates.
- Native form
- Forms single major band in SDS-PAGE
- CAS: 86088-83-7
Specifications
Specifications
| Content And Storage | -20°C |
| Product Type | Bovine Plasma Fibronectin |
| Formulation | In 150mM NaCl, 20mM sodium phosphate buffer, pH 7.3. |
| Quantity | 5 mg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction