Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ AEBSF, Hydrochloride

Catalog No. p-7060015
Click to view available options
Quantity:
50 mg
100 mg
500 mg
1 g

AEBSF, CAS 30827-99-7, is a specific irreversible inhibitor of chymotrypsin, kallikrein, plasmin, thrombin, trypsin, and related thrombolytic enzymes. A nontoxic alternative to PMSF and DFP.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color White to Off-white
Target Proteins chymotrypsin
CAS 30827-99-7
Molecular Formula C8H10FNO2S·HCl
Formula Weight 239.7g/mol
Solubility H2O
Content And Storage Protect from moisture
Form Solid
Quantity 500 mg
Purity >97% by HPLC
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.