Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Leupeptin, Hemisulfate, Synthetic

Catalog No. p-7060017 Shop All MilliporeSigma Products
Click to view available options
Quantity:
10 mg
25 mg

Synthetic version of the microbial-derived inhibitor Leupeptin against trypsin-like and cysteine protease activities.

  • Packaged Under Inert Gas
  • Peptide Sequence: Ac-Leu-Leu-arginal, hemisulfate
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
CAS 103476-89-7
Molecular Formula C20H38N6O4·0.5H2SO4
Formula Weight 475.6g/mol
Solubility H2O
Content And Storage Hygroscopic
Form Solid
Quantity 10 mg
Purity 95% by HPLC (sum of isomers)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.