Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ BSA, Fraction V, Crystalline

Catalog No. p-7078270 Shop All MilliporeSigma Products
Click to view available options
Quantity:
1 g
5 g
25 g

Catalog No. 12-657-1GM

Supplier: MilliporeSigma™ 126571GM

Only null left
Add to Cart
Add to Cart

Prepared by cold ethanol extraction.

  • Crystalline form
  • Contaminants
  • Moisture: ≤5%
  • Assay: ≥99% by Cellulose Acetate Electrophoresis
  • CAS: 9048-46-8

Recommended Storage

2° to 8°C

TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.