Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ BSA, Fraction V, Fatty Acid-Poor, Endotoxin-Free
GSA_VA

Catalog No. p-7078272 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-657-9100GM 100 g
12-657-910GM 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 12-657-9100GM Supplier MilliporeSigma™ Supplier No. 126579100GM
Only null left
Add to Cart
Add to Cart

Designed for use in cell culture systems, animal perfusion studies.

  • Fatty acid poor
  • Endotoxin free
  • Contaminants
  • Ash: ≤2.0%
  • Fatty acids: ≤0.2%
  • Moisture: ≤5%
  • Assay: ≥ 95% by Albumin Electrophoresis
  • CAS: 9048-46-8
  • Color: Pale green

Recommended Storage

2° to 8°C

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 100 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.