Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Calpain Inhibitor VI

Catalog No. 2087455MG Shop All MilliporeSigma Products
Click to view available options
Quantity:
1 mg
5 mg

The Calpain Inhibitor VI, also referenced under CAS 190274-53-4, controls the biological activity of Calpain. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

  • Packaged Under Inert Gas
  • Peptide Sequence: 4-Fluorophenylsulfonyl-Val-Leu-CHO
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color White
Target Proteins μ-calpain
CAS 190274-53-4
Molecular Formula C17H25FN2O4S
Formula Weight 372.5g/mol
Solubility DMSO
Content And Storage Protect from light
Form Lyophilized
Quantity 5 mg
Purity 97% by HPLC
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.