Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Caspase-1 Inhibitor VI

Catalog No. 2187461MG
Click to view available options
Quantity:
1 mg
250 μg

Caspase-1 Inhibitor VI, Z-YVAD-FMK, is a potent, cell-permeable, and irreversible inhibitor of caspase-1, caspase-4, and caspase-5.

  • Peptide Sequence: Z-Tyr-Val-Ala-Asp(OMe)-CH2F*
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color White
Target Proteins caspase-1
Molecular Formula C31H39FN4O9
Formula Weight 630.7g/mol
Solubility DMSO
Content And Storage Protect from moisture
Form Powder
Quantity 1 mg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.