Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ Caspase-3/7 Inhibitor I
Click to view available options
Quantity:
1 mg
Description
- Packaged Under Inert Gas
- Peptide Sequence: 5-[(S)-(+)-2-(Methoxymethyl)pyrrolidino]sulfonylisatin
- Cell Permeable

Specifications
Specifications
Color | Yellow |
Description | 98% by HPLC |
Target Proteins | caspase-3 |
CAS | 220509-74-0 |
Molecular Formula | C14H16N2O5S |
Formula Weight | 324.4g/mol |
Solubility | DMSO or MeOH |
Content And Storage | Protect from light |
Form | Solid |
Quantity | 1 mg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction