Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ CGP-37157
Click to view available options
Quantity:
5 mg
Description
- Packaged Under Inert Gas
- Cell Permeable

Specifications
Specifications
Color | White |
Description | 98% by HPLC |
Target Proteins | Mitochondrial Na+/Ca2+ exchanger |
CAS | 75450-34-9 |
Molecular Formula | C15H11Cl2NOS |
Formula Weight | 324.2g/mol |
Solubility | DMSO |
Content And Storage | Protect from light |
Form | Solid |
Quantity | 5 mg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction