Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ CRAC Channel Inhibitor III, Pyr6
Click to view available options
Quantity:
10 mg
Description
A cell-permeable pyrazole derivative that acts as a selective blocker of Stim1 and Orai1-coupled CRAC (Ca2+ release-activated Ca2+) channel-mediated SOCE (store-operated Ca2+ entry) in RBL-2H3 cells (IC50 = 490nM by Fura-2), while exhibiting much reduced potency against the transient receptor potential cation channel TRPC3-medated ROCE (receptor-operated Ca2+ entry) upon carbachol (Cat. No. 212385) stimulation in TRPC3-expressing HEK293 cells (IC50 = 18.46μM; by Fura-2). A great complement to Pyr2 (Cat. No.203890), Pyr3 (Cat. No. 648490), and Pyr10 (Cat. No. 648494) in Ca2+ signaling studies.

Specifications
Specifications
Color | Light Beige |
Description | 99% by HPLC |
Target Proteins | Orai3 |
Molecular Formula | C17H9F7N4O |
Formula Weight | 418.3g/mol |
Solubility | DMSO |
Content And Storage | Protect from light |
Form | Solid |
Quantity | 10 mg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction