Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ Cyclopamine-KAAD
Shop All MilliporeSigma Products
Click to view available options
Quantity:
100 μg
500 μg
Description
- Packaged Under Inert Gas
- Cell Permeable
Specifications
Specifications
| Color | Pale Yellow |
| Target Proteins | Hh signaling in Shh-light2 assay |
| Molecular Formula | C44H63N3O4 |
| Formula Weight | 698g/mol |
| Solubility | DMSO, EtOH, or MeOH |
| Content And Storage | Protect from light |
| Form | Solid |
| Quantity | 500 μg |
| Purity | 70% by HPLC (sum of two isomers) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction