Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ Cyclopamine, V. californicum
Shop All MilliporeSigma Products
Click to view available options
Quantity:
1 mg
5 mg
Description
A cell-permeable steroidal alkaloid and cholesterol mimic that displays both teratogenic and antitumor activities. It disrupts cholesterol bio-synthesis and specifically antagonizes shh (Sonic Hedgehog) signaling pathway through direct interaction with Smo (smoothened), a distant relative of G-protein-coupled receptors. A valuable tool for studying the involvement of shh signaling in the development of various tumors. Tomatidine (Cat. No. 614350) can be used as a negative control. Also available as a 10mm solution in Ethanol (Cat. No. 239806).
Specifications
Specifications
| Color | Off-white |
| Target Proteins | Hh signaling in Shh-light2 assay |
| CAS | 4449-51-8 |
| Molecular Formula | C27H41NO2 |
| Formula Weight | 411.6g/mol |
| Solubility | DMSO, EtOH, or DMF |
| Content And Storage | Protect from light |
| Form | Solid |
| Quantity | 1 mg |
| Purity | 97% by HPLC |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction