Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ Cyclopiazonic Acid, Penicillium cyclopium
Click to view available options
Quantity:
5 mg
Description
Cell Permeable

Specifications
Specifications
Color | Off-white |
Description | 97% by TLC |
Target Proteins | Ca2+ uptake in cultured arterial musclue cells |
CAS | 18172-33-3 |
Molecular Formula | C20H20N2O3 |
Formula Weight | 336.4g/mol |
Solubility | CHCl3 or DMSO |
Form | Solid |
Quantity | 5 mg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction