Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ D-Amino Acid Oxidase Inhibitor
Click to view available options
Quantity:
50 mg
Description
Packaged Under Inert Gas

Specifications
Specifications
Color | Beige |
Description | 97% by HPLC |
CAS | 39793-31-2 |
Molecular Formula | C7H5NO2S |
Formula Weight | 167.2g/mol |
Solubility | DMSO or 5% NaOH |
Content And Storage | Protect from light |
Form | Solid |
Quantity | 50 mg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction