Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ DUB Inhibitor IV, b-AP15
Click to view available options
Quantity:
10 mg
Description
Packaged Under Inert Gas

Specifications
Specifications
Color | Yellow |
Description | 97% by HPLC |
CAS | 1009817-63-4 |
Molecular Formula | C22H17N3O6 |
Formula Weight | 419.4g/mol |
Solubility | DMSO |
Content And Storage | Protect from light |
Form | Powder |
Quantity | 10 mg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction