Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ Dvl-PDZ Domain Inhibitor, Peptide Pen-N3
Click to view available options
Quantity:
2 mg
Description
- Packaged Under Inert Gas
- Peptide Sequence: Ac-Arg-Gln-Ile-Lys-Ile-Trp-Phe-Gln-Asn-Arg-Arg-Met-Lys-Trp-Lys-Lys-Gly-Gly-Gly-Glu-Ile-Val-Leu-Trp-Ser-Asp-Ile-Pro-NH2

Specifications
Specifications
Color | White |
Formulation | Supplied as a trifluoroacetate salt |
Description | 97% by HPLC |
Molecular Formula | C163H256N48O37S |
Formula Weight | 3512.2g/mol |
Solubility | DMSO |
Content And Storage | Hygroscopic, Protect from light |
Form | Lyophilized |
Quantity | 2 mg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction