Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ E-64 Protease Inhibitor
Shop All MilliporeSigma Products
Click to view available options
Quantity:
1 mg
5 mg
25 mg
Description
Irreversible inhibitor of cysteine proteases. Interacts with the Sn subsites of proteases. Has no action on cysteine residues in other proteins. Inhibits activation-induced programmed cell death and restores defective immune responses in HIV+ donors. Specific active site titrant.

Specifications
Specifications
Color | White |
Target Proteins | cysteine proteases |
CAS | 66701-25-5 |
Molecular Formula | C15H27N5O5 |
Formula Weight | 357.4g/mol |
Solubility | DMSO |
Form | Solid |
Quantity | 1 mg |
Purity | 98% by HPLC |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction